Gene Transcripts
- Gene Symbol
- RAB9BP1
- Entrez Gene ID
- 9366
- Gene Name
- RAB9B, member RAS oncogene family pseudogene 1
- Gene Aliases
- RAB9P1
- Chromosome Location
- Chr. 5: 105099474 - 105100098 on Build GRCh38
- UniGene ID
- Hs.158296
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_000039.2 |
|
1 |
166 |
|
View Full Product Details