Gene Transcripts
- Gene Symbol
- SDHAP1
- Entrez Gene ID
- 255812
- Gene Name
- succinate dehydrogenase complex flavoprotein subunit A pseudogene 1
- Gene Aliases
- SDHAL1, SDHALP1
- Chromosome Location
- Chr. 3: 195959921 - 195990279 on Build GRCh38
- UniGene ID
- Hs.566872
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003264.2 |
|
12, 13 |
1846 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ648899.1 |
- |
|
454 |
|
Chr. - - [-] |
View Full Product Details