Gene Transcripts
- Gene Symbol
- TTTY5
- Entrez Gene ID
- 83863
- Gene Name
- testis-specific transcript, Y-linked 5 (non-protein coding)
- Gene Aliases
- LINC00126, NCRNA00126, TTY5
- Chromosome Location
- Chr. Y: 22296798 - 22298876 on Build GRCh38
- UniGene ID
- Hs.121389
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001541.1 |
|
2 |
1469 |
|
- Gene Symbol
- PRORY
- Entrez Gene ID
- 100533178
- Gene Name
- proline rich, Y-linked
- Gene Aliases
- CYorf17
- Chromosome Location
- Chr. Y: 21381674 - 21454622 on Build GRCh38
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF332236.3 |
- |
2 |
1469 |
|
Chr. Y - 22296794 - 22298876 [-] |
View Full Product Details