Gene Transcripts
- Gene Symbol
- GGNBP1
- Entrez Gene ID
- 449520
- Gene Name
- gametogenetin binding protein 1 (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 6: 33583699 - 33589026 on Build GRCh38
- UniGene ID
- Hs.582973
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028361.1 |
|
4 |
655 |
|
GenBank mRNA |
AY383626.1 |
- |
4 |
616 |
|
AY383627.1 |
- |
4 |
616 |
|
View Full Product Details