Gene Transcripts
- Gene Symbol
- KHDC1
- Entrez Gene ID
- 80759
- Gene Name
- KH homology domain containing 1
- Gene Aliases
- C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4
- Chromosome Location
- Chr. 6: 73241314 - 73310365 on Build GRCh38
- UniGene ID
- Hs.433062
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA540857.1 |
- |
2 |
537 |
|
Chr. 6 - 73275535 - 73310057 [-] |
GenBankmRNA |
BM544101.1 |
- |
2 |
646 |
|
Chr. 6 - 73242071 - 73310149 [-] |
GenBankmRNA |
DW435058.1 |
- |
2 |
222 |
|
Chr. 6 - 73291976 - 73309729 [-] |
GenBankmRNA |
BM760106.1 |
- |
3 |
268 |
|
Chr. 6 - 73275088 - 73310365 [-] |
GenBankmRNA |
BM754816.1 |
- |
3 |
268 |
|
Chr. 6 - 73275049 - 73310365 [-] |
View Full Product Details