Gene Transcripts
- Gene Symbol
- INE2
- Entrez Gene ID
- 8551
- Gene Name
- inactivation escape 2 (non-protein coding)
- Gene Aliases
- NCRNA00011
- Chromosome Location
- Chr. X: 15785716 - 15787589 on Build GRCh38
- UniGene ID
- Hs.653287
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002725.2 |
|
1 |
1076 |
|
GenBank mRNA |
Y10697.1 |
- |
1 |
1072 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY214243.1 |
- |
1 |
129 |
|
Chr. X - 15786230 - 15786614 [-] |
View Full Product Details