Gene Transcripts
- Gene Symbol
- SNORA19
- Entrez Gene ID
- 641451
- Gene Name
- small nucleolar RNA, H/ACA box 19
- Gene Aliases
- ACA19
- Chromosome Location
- Chr. 10: 119060011 - 119060138 on Build GRCh38
- UniGene ID
- Hs.688653
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002917.1 |
|
1 |
61 |
|
GenBank mRNA |
AJ609435.1 |
- |
1 |
61 |
|
View Full Product Details