Gene Transcripts
- Gene Symbol
- ABCC13
- Entrez Gene ID
- 150000
- Gene Name
- ATP binding cassette subfamily C member 13 (pseudogene)
- Gene Aliases
- ABCC13P, C21orf73, PRED6
- Chromosome Location
- Chr. 21: 14273799 - 14301386 on Build GRCh38
- UniGene ID
- Hs.366575
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DN991749.1 |
- |
3 |
375 |
|
Chr. 21 - 14273968 - 14291587 [+] |
GenBankmRNA |
BX101893.1 |
- |
3 |
418 |
|
Chr. 21 - 14273953 - 14291398 [+] |
View Full Product Details