Gene Transcripts

Gene Symbol
KLRA1P
Entrez Gene ID
10748
Gene Name
killer cell lectin like receptor A1, pseudogene
Gene Aliases
KLRA1, KLRAP1, LY49L, Ly-49L, Ly49
Chromosome Location
Chr. 12: 10588478 - 10599835 on Build GRCh38
UniGene ID
Hs.159297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028045.1 7 1784
GenBank mRNA AK310151.1 - 6 1204
CN269037.1 - 1 268

View Full Product Details