Gene Transcripts

Gene Symbol
PPBPP2
Entrez Gene ID
10895
Gene Name
pro-platelet basic protein pseudogene 2
Gene Aliases
PPBPL2, SPBPBP
Chromosome Location
Chr. 4: 74054038 - 74055399 on Build GRCh38
UniGene ID
Hs.3134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026769.1 1 868
GenBank mRNA BC132938.1 - 1 791
BC132940.1 - 1 790
L10403.1 - 1 868

View Full Product Details