Gene Transcripts
- Gene Symbol
- ATP8B5P
- Entrez Gene ID
- 158381
- Gene Name
- ATPase phospholipid transporting 8B5, pseudogene
- Gene Aliases
- FetA
- Chromosome Location
- Chr. 9: 35406755 - 35483029 on Build GRCh38
- UniGene ID
- Hs.575661
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB461154.1 |
- |
7 |
419 |
|
Chr. 9 - 35406789 - 35447316 [+] |
GenBankmRNA |
DB456494.1 |
- |
8 |
490 |
|
Chr. 9 - 35406789 - 35447278 [+] |
View Full Product Details