Gene Transcripts

Gene Symbol
INE1
Entrez Gene ID
8552
Gene Name
inactivation escape 1 (non-protein coding)
Gene Aliases
NCRNA00010
Chromosome Location
Chr. X: 47204848 - 47205861 on Build GRCh38
UniGene ID
Hs.657350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024616.1 1 105
GenBank mRNA BC069671.1 - 1 28
BC069696.1 - 1 27
BC069772.1 - 1 27
DA439607.1 - 1 530
Y10696.1 - 1 32

View Full Product Details