Gene Transcripts
- Gene Symbol
- BPESC1
- Entrez Gene ID
- 60467
- Gene Name
- blepharophimosis, epicanthus inversus and ptosis, candidate 1 (non-protein coding)
- Gene Aliases
- NCRNA00187
- Chromosome Location
- Chr. 3: 139104185 - 139125167 on Build GRCh38
- UniGene ID
- Hs.512673
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000418282.2 |
- |
1 |
319 |
|
Chr. 3 - 139104185 - 139125167 [+] |
View Full Product Details