Gene Transcripts
- Gene Symbol
- C3P1
- Entrez Gene ID
- 388503
- Gene Name
- complement component 3 precursor pseudogene
- Gene Aliases
- CPLP
- Chromosome Location
- Chr. 19: 10041356 - 10074138 on Build GRCh38
- UniGene ID
- Hs.655363
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027300.2 |
|
29 |
3527 |
|
GenBank mRNA |
AK131489.1 |
- |
27 |
3292 |
|
AW827199.1 |
- |
2 |
178 |
|
View Full Product Details