Gene Transcripts
- Gene Symbol
- FMR1-AS1
- Entrez Gene ID
- 100126270
- Gene Name
- FMR1 antisense RNA 1
- Gene Aliases
- ASFMR1, FMR1-AS, FMR1AS, FMR4
- Chromosome Location
- Chr. X: 147909431 - 147911817 on Build GRCh38
- UniGene ID
- Hs.561999
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000596112.5 |
- |
3 |
610 |
|
Chr. X - 147909431 - 147911817 [-] |
GENCODE |
ENST00000594922.5 |
- |
2 |
770 |
|
Chr. X - 147909431 - 147911817 [-] |
GENCODE |
ENST00000598667.1 |
- |
2 |
506 |
|
Chr. X - 147909431 - 147911817 [-] |
View Full Product Details