Gene Transcripts

Gene Symbol
LOH12CR2
Entrez Gene ID
503693
Gene Name
loss of heterozygosity, 12, chromosomal region 2 (non-protein coding)
Gene Aliases
LOH2CR12
Chromosome Location
Chr. 12: 12355408 - 12357067 on Build GRCh38
UniGene ID
Hs.67553
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024061.1 2 1138
GenBank mRNA AY037867.1 - 2 1138


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB049004.1 - 1 200 Chr. 12 - 12355517 - 12355999 [-]

View Full Product Details