Gene Transcripts
- Gene Symbol
- LOH12CR2
- Entrez Gene ID
- 503693
- Gene Name
- loss of heterozygosity, 12, chromosomal region 2 (non-protein coding)
- Gene Aliases
- LOH2CR12
- Chromosome Location
- Chr. 12: 12355408 - 12357067 on Build GRCh38
- UniGene ID
- Hs.67553
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024061.1 |
|
2 |
1420 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AJ707350.1 |
- |
1 |
272 |
|
Chr. 12 - 12355406 - 12355794 [-] |
View Full Product Details