Gene Transcripts
- Gene Symbol
- C21orf62-AS1
- Entrez Gene ID
- 54067
- Gene Name
- C21orf62 antisense RNA 1
- Gene Aliases
- C21orf49
- Chromosome Location
- Chr. 21: 32772100 - 32797707 on Build GRCh38
- UniGene ID
- Hs.54725
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX477211.1 |
- |
4 |
475 |
|
Chr. 21 - 32772162 - 32797168 [+] |
GenBankmRNA |
AV717800.1 |
- |
3 |
329 |
|
Chr. 21 - 32772174 - 32797446 [+] |
View Full Product Details