Gene Transcripts
- Gene Symbol
- MIR98
- Entrez Gene ID
- 407054
- Gene Name
- microRNA 98
- Gene Aliases
- MIRN98, hsa-mir-98, miR-98
- Chromosome Location
- Chr. X: 53556223 - 53556341 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029513.1 |
|
1 |
95 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA362525.1 |
- |
1 |
104 |
|
Chr. X - 53556072 - 53556350 [-] |
View Full Product Details