Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr. 11: 70078302 - 70189545 on Build GRCh38
UniGene ID
Hs.503074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_030691.1 12, 13 1496
XM_006718605.2 XP_006718668.1 12, 13 1491
XM_011545124.2 XP_011543426.1 13, 14 1422
XM_011545126.2 XP_011543428.1 13, 14 1422
XM_011545128.2 XP_011543430.1 12, 13 1356
XM_011545131.2 XP_011543433.1 12, 13 1364
XM_017017957.1 XP_016873446.1 12, 13 1356
GenBank mRNA AK097619.1 - 12, 13 1364
BC033036.2 - 12, 13 1452 4823645

View Full Product Details