Gene Transcripts
- Gene Symbol
- CXADRP3
- Entrez Gene ID
- 440224
- Gene Name
- coxsackie virus and adenovirus receptor pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 18: 14477952 - 14498706 on Build GRCh38
- UniGene ID
- Hs.721675
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024076.1 |
|
2 |
135 |
|
- Gene Symbol
- LOC100287357
- Entrez Gene ID
- 100287357
- Gene Name
- coxsackie virus and adenovirus receptor pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 15: 20803791 - 20804886 on Build GRCh38
- UniGene ID
- Hs.640998
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL834140.1 |
- |
2 |
45 |
|
View Full Product Details