Gene Transcripts
- Gene Symbol
- SNORA9
- Entrez Gene ID
- 677798
- Gene Name
- small nucleolar RNA, H/ACA box 9
- Gene Aliases
- ACA9, SNORA9A
- Chromosome Location
- Chr. 7: 44985378 - 44985510 on Build GRCh38
- UniGene ID
- Hs.744745
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002952.1 |
|
1 |
36 |
|
GenBank mRNA |
AJ609431.1 |
- |
1 |
36 |
|
- Gene Symbol
- SNHG15
- Entrez Gene ID
- 285958
- Gene Name
- small nucleolar RNA host gene 15
- Gene Aliases
- C7orf40, Linc-Myo1g, MYO1GUT
- Chromosome Location
- Chr. 7: 44983028 - 44986660 on Build GRCh38
- UniGene ID
- Hs.25892
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096179.1 |
- |
1 |
1163 |
|
View Full Product Details