Gene Transcripts

Gene Symbol
SNORA14B
Entrez Gene ID
677802
Gene Name
small nucleolar RNA, H/ACA box 14B
Gene Aliases
ACA14b
Chromosome Location
Chr. 1: 235127803 - 235127937 on Build GRCh38
UniGene ID
Hs.744743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002956.1 1 22
GenBank mRNA AJ609457.1 - 1 22


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA248657.1 - 1 271 Chr. 1 - 235122341 - 235128186 [-]
GenBankmRNA DA188431.1 - 1 279 Chr. 1 - 235127661 - 235128193 [-]
GenBankmRNA DA225811.1 - 1 275 Chr. 1 - 235127656 - 235128190 [-]
GenBankmRNA DA115599.1 - 1 111 Chr. 1 - 235113878 - 235128026 [-]

View Full Product Details