Gene Transcripts

Gene Symbol
HAR1B
Entrez Gene ID
768097
Gene Name
highly accelerated region 1B (non-protein coding)
Gene Aliases
HAR1R, LINC00065, NCRNA00065
Chromosome Location
Chr. 20: 63095493 - 63102319 on Build GRCh38
UniGene ID
Hs.635159
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003245.1
2 411
GenBank mRNA DQ860410.1 - LIT3338
HAR1R
2 411


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY011775.1 - 2 401 Chr. 20 - 63095871 - 63102309 [-]
GenBankmRNA HY023235.1 - 2 375 Chr. 20 - 63095892 - 63102283 [-]
GenBankmRNA DB088004.1 - 2 372 Chr. 20 - 63095798 - 63102279 [-]

View Full Product Details