Gene Transcripts
- Gene Symbol
- NOP14-AS1
- Entrez Gene ID
- 317648
- Gene Name
- NOP14 antisense RNA 1
- Gene Aliases
- C4orf10, RES4-24
- Chromosome Location
- Chr. 4: 2935546 - 2951078 on Build GRCh38
- UniGene ID
- Hs.398178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015453.2 |
|
1 |
1261 |
|
GenBank mRNA |
AB000464.1 |
- |
1 |
1261 |
|
AB000466.1 |
- |
1 |
1261 |
|
View Full Product Details