Gene Transcripts
- Gene Symbol
- GGT8P
- Entrez Gene ID
- 645367
- Gene Name
- gamma-glutamyltransferase 8 pseudogene
- Gene Aliases
- GGT1P1
- Chromosome Location
- Chr. 2: 91775342 - 91782127 on Build GRCh38
- UniGene ID
- Hs.650223
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003503.1 |
|
2 |
2142 |
|
GenBank mRNA |
AK055838.1 |
- |
2 |
2142 |
|
- Gene Symbol
- GGT2
- Entrez Gene ID
- 728441
- Gene Name
- gamma-glutamyltransferase 2
- Gene Aliases
- GGT, GGT 2
- Chromosome Location
- Chr. 22: 21207972 - 21282982 on Build GRCh38
- UniGene ID
- Hs.568255
- Species
- Homo sapiens
- Gene Symbol
- LOC102724197
- Entrez Gene ID
- 102724197
- Gene Name
- inactive gamma-glutamyltranspeptidase 2-like
- Gene Aliases
- GGT2
- Chromosome Location
- Chr. 22|NT_187386: 65299 - 86956 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details