Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_024628.1 | 1 | 99 | ||
NR_024629.1 | 1 | 99 | |||
NR_027708.1 | 1 | 99 | |||
NR_027709.1 | 1 | 99 | |||
GenBank mRNA | AF220183.1 | - | 1 | 70 | |
AK314921.1 | - | 1 | 99 | ||
BF724542.1 | - | 1 | 62 |