Gene Transcripts

Gene Symbol
ZFR2
Entrez Gene ID
23217
Gene Name
zinc finger RNA binding protein 2
Gene Aliases
KIAA1086
Chromosome Location
Chr. 19: 3804024 - 3869029 on Build GRCh38
UniGene ID
Hs.65750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027149.1 2 162
GenBank mRNA AL365406.1 - 2 148


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY117144.1 - 2 163 Chr. 19 - 3852357 - 3869030 [-]
GenBankmRNA HY143799.1 - 2 163 Chr. 19 - 3852371 - 3869030 [-]
GenBankmRNA BM690243.1 - 1 136 Chr. 19 - 3852440 - 3855592 [-]
GenBankmRNA BI754220.1 - 2 157 Chr. 19 - 3852009 - 3869024 [-]
GenBankmRNA CN429475.1 - 1 110 Chr. 19 - 3852115 - 3855562 [-]
GenBankmRNA AL536290.3 - 1 100 Chr. 19 - 3851899 - 3855562 [-]
GenBankmRNA CN429473.1 - 3 218 Chr. 19 - 3852362 - 3869023 [-]
GenBankmRNA CN429472.1 - 1 112 Chr. 19 - 3852457 - 3855562 [-]

View Full Product Details