Gene Transcripts
- Gene Symbol
- TPTEP1
- Entrez Gene ID
- 387590
- Gene Name
- transmembrane phosphatase with tensin homology pseudogene 1
- Gene Aliases
- psiTPTE22
- Chromosome Location
- Chr. 22: 16601911 - 16648830 on Build GRCh38
- UniGene ID
- Hs.474116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001591.1 |
|
7, 8 |
1012 |
|
GenBank mRNA |
AY219889.1 |
- |
7, 8 |
1012 |
|
BC107743.1 |
- |
6 |
554 |
|
View Full Product Details