Gene Transcripts
- Gene Symbol
- PPY2P
- Entrez Gene ID
- 23614
- Gene Name
- pancreatic polypeptide 2, pseudogene
- Gene Aliases
- PPY2
- Chromosome Location
- Chr. 17: 28247444 - 28248289 on Build GRCh38
- UniGene ID
- Hs.744962
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002181.1 |
|
1 |
397 |
|
GenBank mRNA |
AF085945.1 |
- |
1 |
292 |
|
AF222903.1 |
- |
1 |
397 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX280431.1 |
- |
1 |
292 |
|
Chr. 17 - 28247549 - 28248216 [+] |
GenBankmRNA |
CR747376.1 |
- |
1 |
292 |
|
Chr. 17 - 28247549 - 28248210 [+] |
View Full Product Details