Gene Transcripts
- Gene Symbol
- LSM5
- Entrez Gene ID
- 23658
- Gene Name
- LSM5 homolog, U6 small nuclear RNA and mRNA degradation associated
- Gene Aliases
- YER146W
- Chromosome Location
- Chr. 7: 32485333 - 32495258 on Build GRCh38
- UniGene ID
- Hs.424908
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024466.1 |
|
1 |
830 |
|
View Full Product Details