Gene Transcripts

Gene Symbol
SEPT7P9
Entrez Gene ID
285961
Gene Name
septin 7 pseudogene 9
Gene Aliases
CDC10L, SEPT7L, bA291L22.2
Chromosome Location
Chr. 10: 38383069 - 38402852 on Build GRCh38
UniGene ID
Hs.406568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027269.1 1, 2 295
GenBank mRNA BC042387.1 - 1, 2 295
BC048143.1 - 1, 2 295
BC171738.1 - 1, 2 261

Gene Symbol
LOC100287497
Entrez Gene ID
100287497
Gene Name
septin 7 pseudogene
Gene Aliases
Chromosome Location
Chr. 1: 223992730 - 224033859 on Build GRCh38
UniGene ID
Hs.568390
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136590.1 1, 2 329
NR_136591.1 1, 2 329
NR_136592.1 3, 4 630
NR_136593.1 1, 2 329
GenBank mRNA AK124970.1 - 1 1627
BX281887.1 - 2, 3 250
DA132394.1 - 1, 2 312
DA229030.1 - 1, 2 312

View Full Product Details