Gene Transcripts

Gene Symbol
YAE1D1
Entrez Gene ID
57002
Gene Name
Yae1 domain containing 1
Gene Aliases
C7orf36, GK003
Chromosome Location
Chr. 7: 39566376 - 39612089 on Build GRCh38
UniGene ID
Hs.72168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282446.1 NP_001269375.1 3 2598
GenBank mRNA AK096857.1 - 1 2434

View Full Product Details