Gene Transcripts

Gene Symbol
NOL8
Entrez Gene ID
55035
Gene Name
nucleolar protein 8
Gene Aliases
C9orf34, NOP132, bA62C3.3, bA62C3.4
Chromosome Location
Chr. 9: 92297358 - 92325594 on Build GRCh38
UniGene ID
Hs.442199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256394.1 NP_001243323.1 4, 5 636
XM_006717169.3 XP_006717232.1 4, 5 551
XM_006717170.3 XP_006717233.1 4, 5 419
XM_006717173.3 XP_006717236.1 4, 5 419
XM_017014877.1 XP_016870366.1 4, 5 419
GenBank mRNA BC146810.1 - 4, 5 392

View Full Product Details