Gene Transcripts
- Gene Symbol
- Mkrn1-ps1
- Entrez Gene ID
- 353327
- Gene Name
- makorin, ring finger protein 1, pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 88837799 - 88839399 on Build GRCm38
- UniGene ID
- Mm.423672
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF494488.1 |
- |
LIT1345 Makorin1-p1 |
1 |
632 |
|
View Full Product Details