Gene Transcripts
- Gene Symbol
- HNRNPL
- Entrez Gene ID
- 3191
- Gene Name
- heterogeneous nuclear ribonucleoprotein L
- Gene Aliases
- HNRPL, P/OKcl.14, hnRNP-L
- Chromosome Location
- Chr. 19: 38836378 - 38852339 on Build GRCh38
- UniGene ID
- Hs.644906
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY927544.1 |
- |
LIT3550 AY927544 |
2 |
325 |
|
View Full Product Details