Gene Transcripts
- Gene Symbol
- BPIFB6
- Entrez Gene ID
- 128859
- Gene Name
- BPI fold containing family B member 6
- Gene Aliases
- BPIL3, LPLUNC6
- Chromosome Location
- Chr. 20: 33031648 - 33044108 on Build GRCh38
- UniGene ID
- Hs.375090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY927631.1 |
- |
LIT3615 AY927631 |
1 |
224 |
|
View Full Product Details