Gene Transcripts

Gene Symbol
NPSR1-AS1
Entrez Gene ID
404744
Gene Name
NPSR1 antisense RNA 1
Gene Aliases
AAA1
Chromosome Location
Chr. 7: 34346512 - 34834331 on Build GRCh38
UniGene ID
Hs.487951
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY312367.1 - LIT3327
AAA1
5 561
U85653.1 -
5 524

View Full Product Details