Gene Transcripts
- Gene Symbol
- PTEN
- Entrez Gene ID
- 5728
- Gene Name
- phosphatase and tensin homolog
- Gene Aliases
- 10q23del, BZS, CWS1, DEC, GLM2, MHAM, MMAC1, PTEN1, TEP1
- Chromosome Location
- Chr. 10: 87863438 - 87971930 on Build GRCh38
- UniGene ID
- Hs.500466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124525.1 |
- |
1 |
89 |
|
View Full Product Details