Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010852.3 | NP_001010852.2 | 6 | 10370 | |
XM_017010266.1 | XP_016865755.1 | 6 | 12072 | ||
GenBank mRNA | AK055705.1 | - | 1 | 1514 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY151725.1 | - | 1 | 90 | Chr. 6 - 123071724 - 123072187 [+] |