Gene Transcripts
- Gene Symbol
- MOP-1
- Entrez Gene ID
- 643616
- Gene Name
- MOP-1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.679418
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB014771.1 |
- |
1 |
233 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CU456651.1 |
- |
1 |
322 |
|
Chr. 4 - 81468836 - 81469398 [-] |
View Full Product Details