Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001261823.1 | NP_001248752.1 | 10 | 2354 | |
NM_003599.3 | NP_003590.1 | 11 | 2445 | ||
NM_181356.2 | NP_852001.1 | 13 | 2630 | ||
GenBank mRNA | AK092340.1 | - | 1 | 713 |