Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321265.1 | NP_001308194.1 | 12 | 6665 | |
NM_001321266.1 | NP_001308195.1 | 11 | 6587 | ||
NM_001321267.1 | NP_001308196.1 | 14 | 6888 | ||
NM_138799.3 | NP_620154.2 | 13 | 6811 | ||
NR_135598.1 | 14 | 6863 | |||
NR_135599.1 | 13 | 7130 | |||
NR_135600.1 | 13 | 6717 | |||
NR_135601.1 | 11 | 6506 | |||
NR_135602.1 | 12 | 6678 | |||
NR_135603.1 | 14 | 6904 | |||
NR_135604.1 | 13 | 6827 | |||
NR_135605.1 | 12 | 6639 | |||
NR_135606.1 | 13 | 6730 | |||
GenBank mRNA | AF052107.1 | - | 1 | 680 |