Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321265.1 | NP_001308194.1 | 12 | 6479 | |
NM_001321266.1 | NP_001308195.1 | 11 | 6401 | ||
NM_001321267.1 | NP_001308196.1 | 14 | 6702 | ||
NM_138799.3 | NP_620154.2 | 13 | 6625 | ||
NR_135598.1 | 14 | 6677 | |||
NR_135599.1 | 13 | 6944 | |||
NR_135600.1 | 13 | 6531 | |||
NR_135601.1 | 11 | 6320 | |||
NR_135602.1 | 12 | 6492 | |||
NR_135603.1 | 14 | 6718 | |||
NR_135604.1 | 13 | 6641 | |||
NR_135605.1 | 12 | 6453 | |||
NR_135606.1 | 13 | 6544 | |||
GenBank mRNA | AF052107.1 | - | 1 | 494 |