Gene Transcripts

Gene Symbol
PROCA1
Entrez Gene ID
147011
Gene Name
protein interacting with cyclin A1
Gene Aliases
Chromosome Location
Chr. 17: 28703197 - 28712065 on Build GRCh38
UniGene ID
Hs.207471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304952.1 NP_001291881.1 2 295
NM_001304953.1 NP_001291882.1 2 412
XM_005257927.4 XP_005257984.1 2 564
XM_006721723.3 XP_006721786.1 2 570
XM_011524398.2 XP_011522700.1 2 294
XM_011524401.1 XP_011522703.1 1 74
GenBank mRNA AK301868.1 - 2 292
AL137531.1 - 1 757
AY601916.1 - 2 320
CD300282.1 - 2 187
DB038438.1 - 2 319


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY021826.1 - 2 390 Chr. 17 - 28704799 - 28711884 [-]
GenBankmRNA DC395780.1 - 2 292 Chr. 17 - 28704188 - 28711086 [-]
GenBankmRNA DB049505.1 - 2 319 Chr. 17 - 28704531 - 28711091 [-]
GenBankmRNA AA398001.1 - 2 158 Chr. 17 - 28704116 - 28710950 [-]
GenBankmRNA HY020995.1 - 2 390 Chr. 17 - 28704726 - 28711884 [-]
GenBankmRNA DB032476.1 - 2 315 Chr. 17 - 28704706 - 28711087 [-]
GenBankmRNA DB028100.1 - 2 315 Chr. 17 - 28704735 - 28711087 [-]
GenBankmRNA BP369255.1 - 2 293 Chr. 17 - 28704304 - 28711086 [-]

View Full Product Details