Gene Transcripts

Gene Symbol
SLC9A3
Entrez Gene ID
6550
Gene Name
solute carrier family 9 member A3
Gene Aliases
DIAR8, NHE-3, NHE3
Chromosome Location
Chr. 5: 471986 - 524434 on Build GRCh38
UniGene ID
Hs.658120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284351.2 NP_001271280.1 17 3349
NM_004174.3 NP_004165.2 17 3376


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY148765.1 - 1 318 Chr. 5 - 472477 - 472935 [-]
GenBankmRNA HY126567.1 - 1 347 Chr. 5 - 472497 - 472965 [-]
GenBankmRNA DA266131.1 - 1 415 Chr. 5 - 472492 - 473034 [-]
GenBankmRNA BI913040.1 - 1 375 Chr. 5 - 472272 - 472994 [-]
GenBankmRNA DA937435.1 - 1 469 Chr. 5 - 472526 - 473088 [-]
GenBankmRNA DA632556.1 - 1 469 Chr. 5 - 472524 - 473088 [-]
GenBankmRNA DA648449.1 - 1 388 Chr. 5 - 472451 - 473007 [-]
GenBankmRNA DA719792.1 - 1 417 Chr. 5 - 472546 - 473036 [-]
GenBankmRNA DA478547.1 - 1 417 Chr. 5 - 472469 - 473036 [-]
GenBankmRNA HY090554.1 - 1 349 Chr. 5 - 472513 - 472967 [-]
GenBankmRNA AU131485.1 - 1 444 Chr. 5 - 472366 - 473063 [-]
GenBankmRNA DB035467.1 - 1 388 Chr. 5 - 472456 - 473007 [-]
GenBankmRNA AK023178.1 - 1 444 Chr. 5 - 470511 - 473063 [-]
GenBankmRNA BI546950.1 - 1 342 Chr. 5 - 472230 - 472956 [-]
GenBankmRNA HY111800.1 - 1 349 Chr. 5 - 472473 - 472968 [-]
GenBankmRNA DA528565.1 - 1 348 Chr. 5 - 472429 - 472967 [-]
GenBankmRNA HY088099.1 - 1 338 Chr. 5 - 472539 - 472956 [-]
GenBankmRNA HY083636.1 - 1 349 Chr. 5 - 472509 - 472967 [-]
GenBankmRNA DA752418.1 - 1 469 Chr. 5 - 472527 - 473088 [-]
GenBankmRNA DA149478.1 - 1 393 Chr. 5 - 472439 - 473012 [-]
GenBankmRNA BG714038.1 - 1 358 Chr. 5 - 472354 - 472960 [-]
GenBankmRNA BU159298.1 - 355 Chr. - - [-]
GenBankmRNA DA696147.1 - 1 388 Chr. 5 - 472460 - 473007 [-]
GenBankmRNA HY119348.1 - 1 318 Chr. 5 - 472432 - 472935 [-]
GenBankmRNA DA786913.1 - 1 393 Chr. 5 - 472440 - 473012 [-]
GenBankmRNA CR988268.1 - 1 406 Chr. 5 - 472422 - 473025 [-]
GenBankmRNA DA616346.1 - 1 456 Chr. 5 - 472499 - 473075 [-]
GenBankmRNA DA297367.1 - 1 368 Chr. 5 - 472412 - 472986 [-]
GenBankmRNA DA048154.1 - 1 417 Chr. 5 - 472501 - 473036 [-]
GenBankmRNA BI917212.1 - 1 333 Chr. 5 - 472354 - 472952 [-]
GenBankmRNA DA525226.1 - 1 358 Chr. 5 - 472418 - 472977 [-]
GenBankmRNA DA478165.1 - 1 456 Chr. 5 - 472519 - 473075 [-]
GenBankmRNA BM807725.1 - 417 Chr. - - [-]
GenBankmRNA DA770151.1 - 1 346 Chr. 5 - 472394 - 472965 [-]
GenBankmRNA DA605548.1 - 1 388 Chr. 5 - 472382 - 473007 [-]
GenBankmRNA DN999780.1 - 1 441 Chr. 5 - 472414 - 473052 [-]
GenBankmRNA DA377199.1 - 1 417 Chr. 5 - 472472 - 473036 [-]
GenBankmRNA DA886782.1 - 1 388 Chr. 5 - 472460 - 473007 [-]
GenBankmRNA DA567671.1 - 1 394 Chr. 5 - 472466 - 473013 [-]
GenBankmRNA HY163438.1 - 1 409 Chr. 5 - 472538 - 473026 [-]
GenBankmRNA DA454975.1 - 1 456 Chr. 5 - 472546 - 473075 [-]
GenBankmRNA DA783068.1 - 1 458 Chr. 5 - 472500 - 473075 [-]
GenBankmRNA DA705860.1 - 1 346 Chr. 5 - 472434 - 472965 [-]
GenBankmRNA CN360410.1 - 1 274 Chr. 5 - 472233 - 472884 [-]
GenBankmRNA BM450275.1 - 1 312 Chr. 5 - 471984 - 472931 [-]
GenBankmRNA HY177673.1 - 1 349 Chr. 5 - 472537 - 472968 [-]
GenBankmRNA BI917207.1 - 1 287 Chr. 5 - 472340 - 472901 [-]
GenBankmRNA DA559396.1 - 1 467 Chr. 5 - 472547 - 473075 [-]
GenBankmRNA HY118290.1 - 1 349 Chr. 5 - 472416 - 472967 [-]
GenBankmRNA HY100748.1 - 1 338 Chr. 5 - 472509 - 472956 [-]
GenBankmRNA DA579043.1 - 1 348 Chr. 5 - 472408 - 472967 [-]
GenBankmRNA DA699271.1 - 1 417 Chr. 5 - 472502 - 473036 [-]
GenBankmRNA DA635333.1 - 1 459 Chr. 5 - 472510 - 473077 [-]
GenBankmRNA AL556583.3 - 1 430 Chr. 5 - 472180 - 473049 [-]

View Full Product Details