Gene Transcripts
- Gene Symbol
- MKRN9P
- Entrez Gene ID
- 400058
- Gene Name
- makorin ring finger protein 9, pseudogene
- Gene Aliases
- MKRN5, MKRN9, MKRNP6, RNF65, ZNF127L3
- Chromosome Location
- Chr. 12: 87782885 - 87784711 on Build GRCh38
- UniGene ID
- Hs.572212
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC067894.1 |
- |
1 |
868 |
|
View Full Product Details