Gene Transcripts

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
Chromosome Location
Chr. 4: 76011180 - 76112802 on Build GRCh38
UniGene ID
Hs.24976
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017008206.1 XP_016863695.1 10 2739
XM_017008207.1 XP_016863696.1 9 2709
XM_017008208.1 XP_016863697.1 12 3252
GenBank mRNA AK129914.1 - 1 425

View Full Product Details