Gene Transcripts
- Gene Symbol
- TARP
- Entrez Gene ID
- 445347
- Gene Name
- TCR gamma alternate reading frame protein
- Gene Aliases
- CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
- Chromosome Location
- Chr. 7: 38239577 - 38276741 on Build GRCh38
- UniGene ID
- Hs.534032
- Species
- Homo sapiens
- Gene Symbol
- TRGV9
- Entrez Gene ID
- 6983
- Gene Name
- T cell receptor gamma variable 9
- Gene Aliases
- TCRGV9, TRGC1, V2
- Chromosome Location
- Chr. 7: 38317017 - 38317492 on Build GRCh38
- UniGene ID
- Hs.534032
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC072387.1 |
- |
1 |
262 |
|
BC072396.1 |
- |
3 |
685 |
|
M16768.1 |
- |
2 |
460 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF085845.1 |
- |
1 |
128 |
|
Chr. 7 - 38316590 - 38317202 [-] |
View Full Product Details