Gene Transcripts

Gene Symbol
KIN
Entrez Gene ID
22944
Gene Name
Kin17 DNA and RNA binding protein
Gene Aliases
BTCD, KIN17, Rts2
Chromosome Location
Chr. 10: 7750962 - 7788027 on Build GRCh38
UniGene ID
Hs.397918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012311.3 NP_036443.1 13 4561
NR_045609.1 14 2136
GenBank mRNA BC037913.2 - 1 929

View Full Product Details